missing translation for 'onlineSavingsMsg'
Learn More

Varenicline Hydrochloride, MedChemExpress

Product Code. 16494246
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494246

Brand: Medchem Express HY10020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Varenicline Hydrochloride (CP 526555 hydrochloride) is a high affinity, selective α4β2 nicotine acetylcholine receptor (nAChR) partial agonist and full α7 nAChR agonist. Varenicline Hydrochloride is also a potent partial agonist of α6β2 nAChR in striatum of rats with a Ki value of 0.12 nM.

Varenicline Hydrochloride (CP 526555 hydrochloride) is a high affinity, selective α4β2 nicotine acetylcholine receptor (nAChR) partial agonist and full α7 nAChR agonist. Varenicline Hydrochloride is also a potent partial agonist of α6β2 nAChR in striatum of rats with a Ki value of 0.12 nM.

Specifications

Chemical Name or Material Varenicline Hydrochloride
CAS 230615-23-3
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C13H14ClN3
Quantity 100 mg
Synonym CP 526555 hydrochloride
Solubility Information H2O : 50 mg/mL (201.84 mM; Need ultrasonic) ∣DMSO : ≥ 2.5 mg/mL (10.09 mM)
SMILES C1(C2CC3CNC2)=C3C=C(N=CC=N4)C4=C1.Cl
Molecular Weight (g/mol) 247.72
Formula Weight 247.72
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.