missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Varicella Zoster Virus Glycoprotein E Antibody (06) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26889-100ug
This item is not returnable.
View return policy
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Varicella Zoster Virus Glycoprotein E | |
| Monoclonal | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| 06 | |
| Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 | |
| Envelope glycoprotein E, GE, ORF68 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction