missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Mineralocorticoid R/NR3C2 Antibody (NR3C2/4900), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26820-20ug
This item is not returnable.
View return policy
Description
Mineralocorticoid R/NR3C2 Monoclonal antibody specifically detects Mineralocorticoid R/NR3C2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Mineralocorticoid R/NR3C2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
| Recombinant fragment (around aa601-673) of human Mineralocorticoid R/NR3C2 protein | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| NR3C2/4900 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4306 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction