missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Varicella Zoster Virus Glycoprotein E Antibody (09) [Alexa Fluor™ 700], Novus Biologicals™

Product Code. 30063148 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30063148 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30063148 Supplier Novus Biologicals™ Supplier No. NBP326914AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot,Sandwich ELISA Detection
TRUSTED_SUSTAINABILITY

Specifications

Antigen Varicella Zoster Virus Glycoprotein E
Applications ELISA, Western Blot, Sandwich ELISA Detection
Classification Monoclonal
Clone 09
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias Envelope glycoprotein E, GE, ORF68
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.