Learn More
Varicella Zoster Virus Glycoprotein E Antibody (144), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-31977-20ug
 View more versions of this product
Product Code. 30116296
  • € 214.00 / 20µg
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Varicella Zoster Virus Glycoprotein E
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Protein A purified
RUO
Virus
Purified
ELISA, Western Blot
144
Western Blot 0.1-1 μg/mL, ELISA 0.1-0.2 μg/mL
Envelope glycoprotein E, GE, ORF68
This antibody was obtained from a rabbit immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538)
20 μg
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.