missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Varicella Zoster Virus Glycoprotein E Antibody (15) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26891MFV500
This item is not returnable.
View return policy
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Varicella Zoster Virus Glycoprotein E | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| Envelope glycoprotein E, GE, ORF68 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| 15 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction