missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VASN Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110822
This item is not returnable.
View return policy
Description
Immunogen sequence: LDVSNLSLQA LPGDLSGLFP RLRLLAAARN PFNCVCPLSW FGPWVRESHV TLASPEETRC HFPPKNAGRL LLELDYADFG CPATTTTATV PTTRPVVREP TALSSSLAPT Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 90%.
The VASN gene encodes the protein vasorin, a type I transmembrane protein involved in cellular signaling, vascular development, and tissue repair. Vasorin is characterized by its expression in various tissues, including vascular endothelial cells and smooth muscle cells, where it functions to modulate cell proliferation and differentiation. It plays a significant role in the regulation of vascular remodeling and homeostasis by interacting with signaling molecules like transforming growth factor-beta (TGF-β), thereby influencing processes such as angiogenesis and wound healing. Vasorin is implicated in inhibiting TGF-β signaling pathways, which are crucial for maintaining vascular integrity and preventing excessive fibrosis during tissue repair. Dysregulation of VASN expression can contribute to pathological conditions, including cardiovascular diseases and cancer, where abnormal vascular remodeling and cell proliferation are evident. Research into vasorin continues to explore its functional mechanisms and potential therapeutic applications in diseases characterized by vascular dysregulation and tissue remodeling.
Specifications
| VASN | |
| Polyclonal | |
| Unconjugated | |
| VASN | |
| 2610528G05Rik; Atia; Protein slit-like 2; Slitl2; Slit-like 2; UNQ314/PRO357/PRO1282; VASN; Vasorin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114990 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6EMK4 | |
| VASN | |
| Recombinant Protein corresonding to Human VASN. Recombinant protein control fragment (Product #RP-108312). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction