Learn More
B Braun™ Vasofix™ Safety Shielded IV Catheter with Injection Port
Product Code. 10323419
Change view
Click to view available options
Quantity:
50/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10323419 50/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10323419 Supplier B Braun™ Supplier No. 4269071S01
  • ONLINE EXCLUSIVE PRICE
    € 1.65
    € 1.61 / Pack of 1
    Save € 0.04
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protects you and your healthcare team against needlestick injuries and related infections.
  • IV catheter material of polyurethane
  • Integrated port allowing injections without an extra needle or risk of needlestick injuries
  • Sharp backcut bevel for minimal puncture trauma
  • Transparent catheter segments provide confirmation of successful catheter placement when withdrawing the needle
  • Latex- and PVC-free
  • According to ISO 10.555-1/5
  • Color coded yellow
  • Size of 24 G, diameter of 0.7 x 19 mm
  • Box of 50 pieces
Specifications
Product Type IV Catheter
Color Yellow
Needle Gauge 24 G
Certifications/Compliance ISO-Standard 10.555-1/5
For Use With (Application) Needlestick injury protection
Length (Metric) Needle 19 mm
Quantity 50/Pk.