Learn More
Vasohibin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-82944
 View more versions of this product
Product Code. 18701304
  • ONLINE EXCLUSIVE PRICE
    € 539.00
    € 509.25 / 0.10mL
    Save € 29.75
    5% Off
Description
Vasohibin Polyclonal specifically detects Vasohibin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Affinity Purified
IgG
VASH1
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.