Learn More
Vasohibin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-82944
This item is not returnable.
View return policy
Description
Vasohibin Polyclonal specifically detects Vasohibin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| Affinity Purified | |
| IgG |
| VASH1 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |