missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vasorin/SLIT-like 2 Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18471JF669
This item is not returnable.
View return policy
Description
Vasorin/SLIT-like 2 Polyclonal antibody specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Vasorin/SLIT-like 2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 669 | |
| Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin | |
| Recognizes region between residues Between between 623 and 673.. | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 114990 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction