Learn More
Vasorin/SLIT-like 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80741
This item is not returnable.
View return policy
Description
Vasorin/SLIT-like 2 Polyclonal specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| Affinity Purified | |
| RUO |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| 0.1 mL | |
| IgG |