Learn More
Vasorin/SLIT-like 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-80741
 View more versions of this product
Product Code. 18705853
  • € 572.00 / 0.10mL
Description
Vasorin/SLIT-like 2 Polyclonal specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Affinity Purified
RUO
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
0.1 mL