missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASP Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41363B
This item is not returnable.
View return policy
Description
VASP Polyclonal antibody specifically detects VASP in Human, Mouse samples. It is validated for Western Blot
Specifications
| VASP | |
| Polyclonal | |
| Western Blot | |
| vasodilator-stimulated phosphoprotein | |
| The epitope is recognized to a region between residue 330 to 380 of human Vasodilator-stimulated phosphoprotein using the numbering given in entry P50552 (GeneID 7408). | |
| 0.1 mL | |
| Phospho Specific | |
| 7408 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction