missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASP Antibody (OTI4D6), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74830APCCY7
This item is not returnable.
View return policy
Description
VASP Monoclonal specifically detects VASP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| VASP | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| VASP | |
| Full length recombinant protein of human VASP (NP_003361) produced in 293T Cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI4D6 | |
| Flow Cytometry | |
| vasodilator-stimulated phosphoprotein | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific | |
| 7408.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction