Learn More
Invitrogen™ VASP Monoclonal Antibody (M277)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA547684
Product Code. 17867018
  • € 564.00 / 100µL
Description
This antibody detects 46 and 50 kDa* proteins corresponding to the apparent molecular mass of VASP on SDS-PAGE immunoblots of human A431, HeLa, and HUVEC.
Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also be involved in the intracellular signaling pathways that regulate integrin-extracellular matrix interactions. VASP is regulated by the cyclic nucleotide-dependent kinases PKA and PKG.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VASP
Monoclonal
0.0625 mg/mL
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
P50552
VASP
Clone (M277) was generated from a recombinant protein that includes amino acids from the C-terminal region of human VASP.
100 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry
M277
Unconjugated
VASP
Enah/Vasp-like; I79_012660; Unknown (protein for MGC:127789); vasodilator stimulated phosphoprotein; vasodilator-stimulated phosphoprotein; vasodilator-stimulated phosphoprotein (VASP); VASP
Mouse
Protein A
RUO
7408
-20°C, Avoid Freeze/Thaw Cycles