missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASP Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502473
This item is not returnable.
View return policy
Description
Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also be involved in the intracellular signaling pathways that regulate integrin-extracellular matrix interactions. VASP is regulated by the cyclic nucleotide-dependent kinases PKA and PKG.Specifications
| VASP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50552 | |
| VASP | |
| Full length human recombit protein of human VASP produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1H8 | |
| Unconjugated | |
| VASP | |
| vasodilator stimulated phosphoprotein | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7408 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction