missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VASP Polyclonal Antibody
GREENER_CHOICE

Product Code. 16316924
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16316924 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16316924 Supplier Invitrogen™ Supplier No. PA578219

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, Neuro2A, C8D30, Raw264.7, C2C12, Rat-2 Predicted Reactivity: Dog (88%), Bovine (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also be involved in the intracellular signaling pathways that regulate integrin-extracellular matrix interactions. VASP is regulated by the cyclic nucleotide-dependent kinases PKA and PKG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VASP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene VASP
Gene Accession No. P50552, P70460
Gene Alias Enah/Vasp-like; I79_012660; Unknown (protein for MGC:127789); vasodilator stimulated phosphoprotein; vasodilator-stimulated phosphoprotein; vasodilator-stimulated phosphoprotein (VASP); VASP
Gene Symbols VASP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human VASP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22323, 361517, 7408
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.