missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAT1L Antibody (OTI1H3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74831JF646
This item is not returnable.
View return policy
Description
VAT1L Monoclonal specifically detects VAT1L in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| VAT1L | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VAT1L | |
| Full length recombinant protein of human VAT1L (NP_065978) produced in HEK293T cells. | |
| 0.1 mL | |
| 57687.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1H3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| KIAA1576synaptic vesicle membrane protein VAT-1 homolog-like, vesicle amine transport protein 1 homolog (T. californica)-like, vesicle amine transport protein 1 homolog-like (T. californica) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction