missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAT1L (KIAA1576) Mouse anti-Human, Mouse, Rat, Clone: OTI1H3, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501107S
This item is not returnable.
View return policy
Description
VAT1L (KIAA1576) Monoclonal antibody specifically detects VAT1L (KIAA1576) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western BlotSpecifications
| VAT1L (KIAA1576) | |
| Monoclonal | |
| 0.62 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q80TB8, Q9HCJ6 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 270097, 361414, 57687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| OTI1H3 | |
| Unconjugated | |
| VAT1L | |
| 9430073I07; AI427515; Kiaa1576; mKIAA1576; Synaptic vesicle membrane protein VAT-1 homolog-like; Vat1l; vesicle amine transport 1-like; vesicle amine transport protein 1 homolog (T. californica)-like; vesicle amine transport protein 1 homolog-like (T. californica) | |
| Full length human recombinant protein of human VAT1L (NP_065978) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction