missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAV3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121509
This item is not returnable.
View return policy
Description
VAV3 Polyclonal Antibody for Western Blot, IHC (P)
This gene is a member of the VAV gene family. The VAV proteins are guanine nucleotide exchange factors (GEFs) for Rho family GTPases that activate pathways leading to actin cytoskeletal rearrangements and transcriptional alterations. This gene product acts as a GEF preferentially for RhoG, RhoA, and to a lesser extent, RAC1, and it associates maximally with the nucleotide-free states of these GTPases. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| VAV3 | |
| Polyclonal | |
| Unconjugated | |
| VAV3 | |
| A530094I06Rik; AA986410; FLJ40431; guanine nucleotide exchange factor VAV3; Idd18.1; RGD1565941; vav 3 guanine nucleotide exchange factor; vav 3 oncogene; vav guanine nucleotide exchange factor 3; Vav3; VAV-3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10451 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9UKW4 | |
| VAV3 | |
| Recombinant protein Vav 3 Oncogene. The antigen corresponds to amino acid range 183-380 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction