missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VBP1 Monoclonal Antibody (OTI1D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504893S
This item is not returnable.
View return policy
Description
VBP1 Monoclonal antibody specifically detects VBP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| VBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7TNY6, Q8BMP6, Q9H3P7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 170760, 289312, 64746 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D1 | |
| Unconjugated | |
| Acbd3 | |
| EGK_21124; HIBBJ46; LOC703257; PFD3; Pfdn3; phosphatidylinositol-binding clathrin assembly protein; prefoldin 3; prefoldin subunit 3; prefoldin subunit 3; phosphatidylinositol-binding clathrin assembly protein; RP13-228J13.4; VBP1; VBP-1; VHL binding protein 1; VHL binding protein-1; VHL-binding protein 1; von Hippel-Lindau binding protein 1; von Hippel-Lindau-binding protein 1 | |
| Full length human recombinant protein of human ACBD3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction