missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VBP1 Monoclonal Antibody (OTI1D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504893
This item is not returnable.
View return policy
Description
The protein encoded by VBP1 interacts with the Von Hippel-Lindau protein to form an intracellular complex. The encoded protein functions as a chaperone protein, and may play a role in the transport of the Von Hippel-Lindau protein from the perinuclear granules to the nucleus or cytoplasm. Alternative splicing and the use of alternate transcription start sites results in multiple transcript variants encoding different protein isoforms.Specifications
| VBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P61758 | |
| VBP1 | |
| Full length human recombit protein of human VBP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D1 | |
| Unconjugated | |
| VBP1 | |
| HIBBJ46, PFD3, PFDN3, VBP-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7411 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction