missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VBP1 Monoclonal Antibody (OTI2G2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526012
This item is not returnable.
View return policy
Description
VBP1 Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by VBP1 interacts with the Von Hippel-Lindau protein to form an intracellular complex. The encoded protein functions as a chaperone protein, and may play a role in the transport of the Von Hippel-Lindau protein from the perinuclear granules to the nucleus or cytoplasm. Alternative splicing and the use of alternate transcription start sites results in multiple transcript variants encoding different protein isoforms.
Specifications
| VBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61758, P61759 | |
| Vbp1 | |
| Full length human recombinant protein of VBP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G2 | |
| Unconjugated | |
| Vbp1 | |
| EGK_21124; HIBBJ46; LOC703257; PFD3; Pfdn3; phosphatidylinositol-binding clathrin assembly protein; prefoldin 3; prefoldin subunit 3; prefoldin subunit 3; phosphatidylinositol-binding clathrin assembly protein; RP13-228J13.4; VBP1; VBP-1; VHL binding protein 1; VHL binding protein-1; VHL-binding protein 1; von Hippel-Lindau binding protein 1; von Hippel-Lindau-binding protein 1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 22327, 481089, 681825, 7411 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction