missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1/CD106 Antibody (B-K9), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47864PEATT594
This item is not returnable.
View return policy
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications
| VCAM-1/CD106 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| VCAM1 | |
| Activated human umbilical vein endothelial cells (HUVEC) | |
| 0.1 mL | |
| Primary | |
| Recognizes a protein of 110kDa, identified as CD106 (also known as vascular cell adhesion molecule-1 (VCAM-1) and INCAM-100). CD106 is a member of the Ig superfamily of adhesion molecules and is expressed at high levels on cytokine stimulated vascular endothelial cells, and at minimal levels on un-stimulated endothelial cells. It is also present on follicular and inter-follicular dendritic cells of lymph nodes, myoblasts, and some macrophages. CD106 serves as a ligand for leukocyte integrin (VLA-4 or CD49d/CD29) and mediates cell adhesion of leukocytes to activated endothelium. It plays a role in various immunological and inflammatory responses. This monoclonal antibody inhibits the binding of leukocytes to VCAM-1 on stimulated endothelial cells. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| B-K9 | |
| Flow Cytometry | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| 7412.0 | |
| Human, Rat | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction