missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VCAM-1 (CD106) Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143041
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoform a (NP_001069.1) and isoform c (NP_001186763.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
Specifications
| VCAM-1 (CD106) | |
| Polyclonal | |
| Unconjugated | |
| VCAM1 | |
| adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 7412 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P19320 | |
| VCAM1 | |
| Peptide with sequence C-TQIDSPLSGKVRSE (aa 349-362). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction