missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM1 Monoclonal Antibody (OTI3G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502426S
This item is not returnable.
View return policy
Description
VCAM1 Monoclonal antibody specifically detects VCAM1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| VCAM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9ULC4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 28985 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G7 | |
| Unconjugated | |
| MCTS1 | |
| adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
| Human recombinant protein fragment corresponding to amino acids 1-181 of human MCTS1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction