missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VCP Polyclonal Antibody
GREENER_CHOICE

Product Code. 16372845
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16372845 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16372845 Supplier Invitrogen™ Supplier No. PA585940

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VCP Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VCP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene VCP
Gene Accession No. P55072, Q01853, Q7ZU99
Gene Alias 15S Mg(2+)-ATPase p97 subunit; 3110001E05; AAA ATPase p97; ALS14; CDC48; cell division cycle gene CDC48; CMT2Y; epididymis luminal protein 220; epididymis secretory protein Li 70; HEL-220; HEL-S-70; homolog of yeast cdc48; IBMPFD; IBMPFD1; Inv protein; or HEL-S-70; p97; p97 atpase; p97/VCP; Protein CDC48; si:ch211-113n10.2; TER ATPase; TERA; transitional endoplasmic reticulum ATPase; valosin containing protein; valosin-containing protein; Vcp; wu:fd16d05; wu:fj63d11; yeast Cdc48p homolog
Gene Symbols VCP
Host Species Rabbit
Immunogen Synthetic peptide within human VCP aa 757-806.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269523, 327197, 7415
Target Species Human, Mouse, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.