missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCP Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
€ 149.00 - € 439.00
Specifications
| Antigen | VCP |
|---|---|
| Clone | 3G23 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234614
|
Proteintech
82463-1-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237320
|
Proteintech
82463-1-RR-100UL |
100 μL |
€ 439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The VCP antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human VCP. This antibody recognizes human, mouse, rat antigen. The VCP antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| VCP | |
| 1000 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 7415 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 3G23 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| IBMPFD; MGC131997; MGC148092; MGC8560; TERA; p97 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title