Learn More
VDAC2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97576AF594
Product Code. 18049437
  • ONLINE EXCLUSIVE PRICE
    € 438.00
    € 413.70 / 0.10mL
    Save € 24.30
    5% Off
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
VDAC2 Polyclonal specifically detects VDAC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
VDAC2
Polyclonal
50 mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 594
FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2
Produced in rabbits immunized with E. coli-derived Human VDAC2 fragment.
0.1 mL
Lipid and Metabolism
7417
Store at 4°C in the dark.