missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VDAC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100190
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total VDAC3.
The Voltage-Dependent Anion Channel (VDAC) of the outer mitochondrial membrane, a small abundant protein found in all eukaryotic kingdoms, forms a voltage-gated pore when incorporated into planar lipid bilayers. VDAC is also the site of binding of the metabolic enzymes hexokinase and glycerol kinase to the mitochondrion in what may be a significant metabolic regulatory interaction. VDAC3 is involved specifically in translocation of adenine nucleotides through the outer membrane.
Specifications
| VDAC3 | |
| Polyclonal | |
| Unconjugated | |
| VDAC3 | |
| channel protein VDAC3; HD-VDAC3; mitochondrial outer membrane protein; mitochondrial voltage dependent anion channel 3; mVDAC3; outer mitochondrial membrane protein porin 3; rVDAC3; Vdac3; VDAC-3; voltage dependent anion channel 3; voltage-dependent anion channel 3; voltage-dependent anion-selective channel protein 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22335, 7419, 83532 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q60931, Q9R1Z0, Q9Y277 | |
| VDAC3 | |
| A synthesized peptide derived from human VDAC3(Accession Q9Y277), corresponding to amino acid residues D30-T80. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction