missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin Antibody (16B1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00342PCP
This item is not returnable.
View return policy
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human samples. It is validated for Flow Cytometry.
Specifications
| VE-Cadherin | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CDH5 | |
| The immunogen for this antibody is VE Cadherin. | |
| 100 μL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 1003 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 16B1 | |
| Flow Cytometry | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction