missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin Antibody (BV13), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43348AF594
This item is not returnable.
View return policy
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| VE-Cadherin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CDH5 | |
| The immunogen for this antibody was CD144. | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| BV13 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| Rat | |
| Protein A or G purified | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 1003.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction