missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VE-cadherin Recombinant Rabbit Monoclonal Antibody (201), PE
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560416
This item is not returnable.
View return policy
Description
VE-cadherin Recombinant Monoclonal Antibody for Flow
VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.
Specifications
| VE-cadherin | |
| Recombinant Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| P55284 | |
| CDH5 | |
| Recombinant Mouse VE-Cadherin/CD144/CDH5 protein (Met1-Ala592). | |
| 25 Tests | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 201 | |
| PE | |
| CDH5 | |
| 7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12562 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction