missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Statin/EGFL7 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-44320
This item is not returnable.
View return policy
Description
Specifically detects VE-Statin/EGFL7 in Human samples, and it is validated for Peptide ELISA, Western Blotting
VE-Statin/EGFL7 Polyclonal specifically detects VE-Statin/EGFL7 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| VE-Statin/EGFL7 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VE-STATIN, Zneu1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The immunizing peptide represents the N terminus of the mature protein. Reported variants represent identical protein: NP_057299.1, NP_958854.1. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_057299.1 | |
| EGFL7 | |
| Peptide with sequence C-HGDPVSESFVQR, from the internal region of the protein sequence according to NP_057299.1. | |
| 0.1 mg | |
| Angiogenesis | |
| 51162 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction