missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Statin/EGFL7 Antibody (OTI2B5), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74846FR
This item is not returnable.
View return policy
Description
VE-Statin/EGFL7 Monoclonal specifically detects VE-Statin/EGFL7 in Human samples. It is validated for Western Blot.
Specifications
| VE-Statin/EGFL7 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| EGFL7 | |
| Recombinant protein fragment corresponding to amino acids 24-273 of human VE-Statin/EGFL7 (NP_057299) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI2B5 | |
| Western Blot | |
| EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VE-STATIN, Zneu1 | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis | |
| 51162.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction