Learn More
VEGF Antibody (Domantis patent anti-VEGF), Alexa Fluor™ 405, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-27976AF405
Product Code. 30116798
  • € 431.00 / 0.10mL
Description
VEGF Monoclonal antibody specifically detects VEGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VEGF
Monoclonal
Alexa Fluor 405
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
VEGF
0.1 mL
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Cytokines and Growth Factors, Developmental Biology, Glycobiology, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
7422
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
Domantis patent anti-VEGF
50mM Sodium Borate
Human
Protein A purified