missing translation for 'onlineSavingsMsg'
Learn More

VEGF-C Antibody (MM0006-2E65), CoraFluor™ 1, Novus Biologicals™

Product Code. 30146746 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30146746 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30146746 Supplier Novus Biologicals Supplier No. NB11060971CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF-C Monoclonal antibody specifically detects VEGF-C in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF-C
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0006-2E65
Conjugate CoraFluor 1
Formulation PBS
Gene Alias Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Host Species Mouse
Immunogen Human recombinant VEGF-C
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7424
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.