missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-C Antibody (OTI4A1), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74845R
This item is not returnable.
View return policy
Description
VEGF-C Monoclonal specifically detects VEGF-C in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| VEGF-C | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VEGFC | |
| Recombinant protein fragment corresponding to amino acids 112-227 of human VEGF-C (NP_005420) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI4A1 | |
| Western Blot | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
| 7424.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction