missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VEGF-C Antibody (VGX100), Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 920.00
Specifications
| Antigen | VEGF-C |
|---|---|
| Clone | VGX100 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045166
|
Novus Biologicals™
NBP3-28804-1mg |
1 mg |
€ 920.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044522
|
Novus Biologicals™
NBP3-28804-100ug |
100 μg |
€ 326.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046647
|
Novus Biologicals™
NBP3-28804-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF-C Monoclonal antibody specifically detects VEGF-C in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssayTekniske data
| VEGF-C | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
| VEGFC | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| VGX100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P49767 | |
| 7424 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel