missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-D Antibody (106), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89563AF405
This item is not returnable.
View return policy
Description
VEGF-D Monoclonal antibody specifically detects VEGF-D in Human samples. It is validated for ELISA
Specifications
| VEGF-D | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 106 | |
| ELISA | |
| c-fos induced growth factor (vascular endothelial growth factor D), vascular endothelial growth factor D, VEGF-DVEGFDc-Fos-induced growth factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF-D (NP_004460.1; Phe 93-Ser 201) . | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers | |
| 2277 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction