missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-D Antibody (MM0007-7E79), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60973CL1
This item is not returnable.
View return policy
Description
VEGF-D Monoclonal antibody specifically detects VEGF-D in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| VEGF-D | |
| Monoclonal | |
| CoraFluor 1 | |
| c-fos induced growth factor (vascular endothelial growth factor D), vascular endothelial growth factor D, VEGF-DVEGFDc-Fos-induced growth factor | |
| Human recombinant VEGF-D | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers | |
| 2277 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| MM0007-7E79 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction