missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VEGF Receptor 1/2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA116612
This item is not returnable.
View return policy
Description
The target sequence has 100% sequence homology with mouse and rat.
Receptors for VEGF, VEGFB and PGF have a tyrosine-protein kinase activity. The VEGF-kinase ligand/receptor signaling system plays a key role in vascular development and regulation of vascular permeability. VEGF and its high-affinity binding receptors, the tyrosine kinases FLK1 and FLT1, are thought to be important for the development of embryonic vasculature. It has been shown that an alternately spliced form of FLT1 produces a soluble protein, termed sFLT1, which binds vascular endothelial growth factor with high affinity, playing an inhibitory role in angiogenesis.
Specifications
| VEGF Receptor 1/2 | |
| Polyclonal | |
| Unconjugated | |
| FLT1 | |
| 6130401C07; AI323757; CD309; Embryonic receptor kinase 2; Emrk2; Fetal liver kinase 1; fetal liver kinase-1; Flk1; FLK-1; FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2); FLK1 kinase insert domain receptor (VEGF receptor 2); FLT; Flt1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; KDR; kinase insert domain protein receptor; kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); kinase NYK; Krd-1; Ly73; Protein-tyrosine kinase receptor flk-1; sFlt1; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 1; soluble VEGFR2; sVEGFR 1; sVEGFR-2; tyrosine kinase growth factor receptor; tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; vascular endothelial growth factor receptor 1; Vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-1; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; vascular permeability factor receptor; VEGF R1; VEGF receptor-2; VEGFR; VEGFR 1; Vegfr1; VEGFR-1; VEGFR2; Vegfr-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2321, 25589, 3791, 54251 | |
| 4°C, do not freeze | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O08775, P17948, P35968, P53767 | |
| FLT1, KDR | |
| A synthetic peptide made to an internal region of the mouse VEGF Receptor 1 protein (between residues 800-900). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction