missing translation for 'onlineSavingsMsg'
Learn More

VEGF Receptor 1 Recombinant Rabbit Monoclonal Antibody (SY09-09)
GREENER_CHOICE

VEGF Receptor 1 Recombinant Rabbit Monoclonal Antibody (SY09-09)

Brand:  Invitrogen MA532045

Product Code. 16335474

  • € 540.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VEGF Receptor 1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow

TRUSTED_SUSTAINABILITY
Specifications

Specifications

VEGF Receptor 1
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
P17948, P35969, P53767
FLT1
Synthetic peptide within Human VEGF Receptor 1 aa 1-50.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
SY09-09
Unconjugated
FLT1
AI323757; embryonic receptor kinase 2; Emrk2; FLT; FLT1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; i21; sFlt1; soluble VEGFR 1; sVEGFR 1; Tyrosine-protein kinase FRT; tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGF R1; VEGFR1; VEGFR-1
Rabbit
Protein A
RUO
14254, 2321, 54251
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.