missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR-1/FLT-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13687-1-AP-20UL
This item is not returnable.
View return policy
Description
VEGFR 1 is a cell membrane receptor kinase. It has a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.Specifications
| VEGFR-1/FLT-1 | |
| Polyclonal | |
| Unconjugated | |
| FLT1 | |
| FLT, FLT 1, FLT1, FLT-1, Fms like tyrosine kinase 1, FRT, sflt1, Tyrosine protein kinase FRT, VEGF receptor 1, VEGFR 1, VEGFR1, VEGFR-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14254, 2321, 54251 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Flow Cytometry | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P17948, P35969, P53767 | |
| Flt1 | |
| VEGFR-1/FLT-1 Fusion Protein Ag4558 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction