missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR1/Flt-1 Antibody (EWC), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1004JF549
This item is not returnable.
View return policy
Description
VEGFR1/Flt-1 Monoclonal antibody specifically detects VEGFR1/Flt-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| VEGFR1/Flt-1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 | |
| Recombinant human soluble extracellular Flt-1 (D5) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases | |
| 2321 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunofluorescence | |
| EWC | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction