missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR1/Flt-1 Antibody (Z9P4F3*H10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.00 - € 361.00
Specifications
| Antigen | VEGFR1/Flt-1 |
|---|---|
| Clone | Z9P4F3*H10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682847
|
Novus Biologicals
NBP2-50408-0.025ml |
0.025 mL |
€ 201.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664399
|
Novus Biologicals
NBP2-50408 |
0.1 mL |
€ 361.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGFR1/Flt-1 Monoclonal antibody specifically detects VEGFR1/Flt-1 in Zebrafish samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| VEGFR1/Flt-1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases | |
| PBS | |
| 2321 | |
| IgG1 κ | |
| Protein A purified |
| Z9P4F3*H10 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Zebrafish | |
| EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 | |
| Ovalbumin-conjugated synthetic peptide KEIPDDEQSD (C-terminal sequence) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title