missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR3/Flt-4 Antibody (MM0003-7G63), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60968H
This item is not returnable.
View return policy
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| VEGFR3/Flt-4 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| FLT4 | |
| Human VEGFR3/Flt-4 (N-terminal) | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity is shown with VEGFR1 or VEGFR2 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0003-7G63 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Cancer, Hypoxia, Protein Kinase | |
| 2324 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction