missing translation for 'onlineSavingsMsg'
Learn More

Vemurafenib, MedChemExpress

Product Code. 16451207
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16451207

Brand: Medchem Express HY12057

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vemurafenib (PLX4032) is a first-in-class, selective, potent inhibitor of B-RAF kinase, with IC50s of 31 and 48 nM for RAFV600E and c-RAF-1, respectively. Vemurafenib induces cell autophagy.

Vemurafenib (PLX4032) is a first-in-class, selective, potent inhibitor of B-RAF kinase, with IC50s of 31 and 48 nM for RAFV600E and c-RAF-1, respectively. Vemurafenib induces cell autophagy.

Specifications

Chemical Name or Material Vemurafenib
CAS 918504-65-1
Color White
Purity Grade Notes Research
Molecular Formula C23H18ClF2N3O3S
Quantity 50 mg
Synonym PLX4032 RG7204 RO5185426
Solubility Information DMSO : 50 mg/mL (102.06 mM; Need ultrasonic)
SMILES FC1=CC=C(C(F)=C1C(C2=CNC3=NC=C(C=C32)C4=CC=C(C=C4)Cl)=O)NS(CCC)(=O)=O
Molecular Weight (g/mol) 489.92
Formula Weight 489.92
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.