Learn More
Invitrogen™ Venezuelan Equine Encephalitis Virus NSP4 Recombinant Rabbit Monoclonal Antibody (HL2023)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556700
This item is not returnable.
View return policy
Description
Venezuelan Equine Encephalitis Virus NSP4 Recombinant Monoclonal Antibody for Western Blot
Venezuelan Equine Encephalitis Virus (VEEV) is a mosquito-borne virus that can cause severe illness in both humans and animals. The virus is classified as a category B bioterrorism agent by the Centers for Disease Control and Prevention (CDC) due to its potential use as a biological weapon. The NSP4 gene of the VEEV is a non-structural protein that plays a crucial role in viral replication and pathogenesis. NSP4 is a promising target for the development of antiviral drugs and vaccines against VEEV.
Specifications
| Venezuelan Equine Encephalitis Virus NSP4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2023 | |
| Unconjugated | |
| VEEV NSP4 | |
| Recombinant fragment of Venezuelan equine encephalitis virus nsp4. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |