Learn More
B Braun™ Venofix™ Safety Winged IV Needles
Reduce the risks of infection with bloodborne pathogens. B Braun™ Venofix™ Safety Winged IV Needles include a safety mechanism that can be activated with one hand while the needle is still in the patient’s vein.
Brand: B Braun™ 4056510-01
Additional Details : Weight : 0.17100kg
Description
- Easy activation during handling procedure
- Possibility of in-vein activation – no exposure to contaminated needle
- Audible click assures that the safety mechanism is activated
- Once locked, the mechanism stays locked
Specifications
Blue | |
23 G | |
0.65 mm |
Tubing length: 30mm | |
Winged safety mechanism | |
0.49 mL (Residual) |