Learn More
Emma Safety Footwear Venus S2 Low-Profile D-Fit Safety Shoes
Designed with smooth, easy-to-clean contours featuring hygiene advantages.
Brand: Emma Safety Footwear MM71454037
Additional Details : Weight : 0.01000kg
Description
- Full grain leather uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
Specifications
Hydro-Tec sanitized silver lining | |
Non-woven antistatic insole | |
Low-profile Safety Shoe |
37 | |
GripForce™ Easy Twist outsole |